ContactUsLinkComponent
Counting chambers for CSF, BLAUBRAND® Nageotte
Counting chambers for CSF, BLAUBRAND® Nageotte
Catalog # BRND721305
Supplier:  Brand
undefined
Counting chambers for CSF, BLAUBRAND® Nageotte
Catalog # BRND721305
Supplier:  Brand
Specifications
  • Depth of chamber:
    0,5 mm
  • Type:
    Nageotte
  • Description:
    Counting chamber, without clips, double ruling
  • Pk:
    1
Specifications
About this item
This counting chamber is used for counting cells in the cerebro-spinal fluid (CSF) as well as for counting nematodes.
The square area of 100 mm² is subdivided into 40 rectangles with an area of 2,5 mm² each.