Orders LinkContactUsLinkComponent
Pasteur pipettes, Corning®
Pasteur pipettes, Corning®
Supplier:   Corning
Pasteur pipettes, Corning®
Supplier:  Corning
Certificates
Pasteur pipettes, Corning®
Pasteur pipettes, Corning®
3 Options Available Below
Product Details & Documents
Soda-lime or borosilicate glass.
  • Single use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents
  • Features a constriction one inch below the top to facilitate plugging
  • Available in convenient bulk packs
These disposable Pasteur pipettes are ideal for rapid non volumetric transfer work in bacteriology, immunology, haematology and serology studies, as well as blood bank and clinical chemistry procedures.
Documents
Product Family Options
Product Information
  • Ext.Ø
    Length
    Sterile
    Packed
    Availability
    Price
  • 612-5199
    Non plugged, borosilicate glass
    Non plugged, borosilicate glass
    6,5 mm
    146 mm
    -
    200/pack
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 76,70 €
    76,70 €
    1 * 1.000 ST
       76,70 €
  • 612-5200
    Non plugged, borosilicate glass
    Non plugged, borosilicate glass
    6,5 mm
    229 mm
    -
    200/pack
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 85,40 €
    85,40 €
    1 * 1.000 ST
       85,40 €
  • 612-5198
    Non plugged, long tip, soda-lime glass
    Non plugged, long tip, soda-lime glass
    6,5 mm
    330 mm
    -
    100/case
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 120,00 €
    120,00 €
    1 * 100 ST
       120,00 €