ContactUsLinkComponent
Counting chambers, BLAUBRAND® Neubauer Improved
Counting chambers, BLAUBRAND® Neubauer Improved
Catalog # 612-5713
Supplier:  Brand
undefined
Counting chambers, BLAUBRAND® Neubauer Improved
Catalog # 612-5713
Supplier:  Brand
Specifications
  • Type:
    Neubauer Improved
  • Description:
    Counting chamber, without clips, bright-line
  • Pk:
    1
Specifications
About this item
This counting chamber feature the same ruling as improved Neubauer but with a rhodium coated chamber bottom.
  • Rulings engraved into the rhodium layer appear bright under normal microscope settings
  • By altering the contrast, the microscope image can be reversed
  • Rulings can appear brighter or darker, as required