Order Entry
Austria
ContactUsLinkComponent
Sodium hydroxide 0.02 mol/l (0.02 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Sodium hydroxide 0.02 mol/l (0.02 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Catalog # 1.09142.0500
Supplier:  Merck
CAS Number:  
Sodium hydroxide 0.02 mol/l (0.02 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Catalog # 1.09142.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Form:
    liquid
  • Amount-of-substance concentration:
    0.01990 - 0.02002
  • Measurement uncertainty:
    + / - 0.00006
  • Traceability:
    NIST SRM

Specification Test Results

About this item

 :
 :40 g/mol
Density:1 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:1824
ADR:8,II
 :M-Clarity™ quality level = MQ100
EINECS:215-185-5
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548