Order Entry
Austria
ContactUsLinkComponent
 
Nitric acid 1 mol/l (1 N), AVS TITRINORM volumetric solution
Catalog # 30065.291
Supplier:  VWR Chemicals
CAS Number:  
undefined
Nitric acid 1 mol/l (1 N), AVS TITRINORM volumetric solution
Catalog # 30065.291
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.998 - 1.002 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :63.01 g/mol
Density:1.035 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
 :GHS05
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349