Order Entry
Austria
ContactUsLinkComponent
 
 
Nitric acid 1 mol/l (1 N), BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
  7622.9010
 :  Avantor
 :  
 
Nitric acid 1 mol/l (1 N), BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
  7622.9010
 :  Avantor
 :  

 

  • Pack type:
    Plastic container
  • Pk:
    10 L
  • Seq:
    5214

 

 

  • Titer (mol/l):
    0.995 - 1.005

 

 

 :
 :63.01 g/mol
 :1.035 g/cm³ (25 °C)
 :Ambient
 :231-714-2
 :3264
 :8,III
 :GHS05
 :7697-37-2
 :HNO₃
 :MFCD00011349