
Potassium permanganate 0.02 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7722-64-7
:
Formula:KMnO₄
MDL Number:MFCD00011364
:158.03 g/mol
Storage Temperature:Ambient
EINECS:231-760-3
UN:3082
ADR:9,III
:GHS09
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):0.01996 - 0.02004 mol/l
- NIST traceable:Confirmed
Specification Test Results
Frequently Bought Together