Potassium permanganate 0.02 mol/l (0.1 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7722-64-7
:
Formula:KMnO₄
MDL Number:MFCD00011364
:158.03 g/mol
ADR:9,III
:M-Clarity™ quality level = MQ100
EINECS:231-760-3
UN:3082
:GHS09
Storage Temperature:Ambient
Limited Time Sale
Select items below are currently on sale
Related Information
Certificates
Search by Lot NumberMerck Brighter titration Titripur®
Product Literature
ProductPage_109121
Product specification
Frequently Bought Together