

Perchloric acid 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 30111.291
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):0.998 - 1.002 mol/l
- NIST traceable:Confirmed
Specification Test Results
About this item
:
:100.46 g/mol
Storage Temperature:Ambient
EINECS:231-512-4
UN:3264
ADR:8,III
:GHS05
:GHS03
CAS Number:7601-90-3
Formula:HClO₄
MDL Number:MFCD00011325