

Perchloric acid 0.1 mol/l (0.1 N) in anhydrous acetic acid, AVS TITRINORM Reag. Ph. Eur. volumetric solution
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7601-90-3
:
Formula:HClO₄
MDL Number:MFCD00011325
:100.46 g/mol
Boiling Pt:120 °C (1013 hPa)
Density:1.050 g/cm³ (20 °C)
Flash Pt:40 °C (closed cup)
Storage Temperature:Ambient
EINECS:231-512-4
UN:2920
ADR:8 (3),II
:GHS02
:GHS05
Specification Test Results
- Titer (20°C; real value 0.5 % accuracy):0.0998 - 0.1002 mol/l
- NIST traceable:Confirmed
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle Safebreak for liquids DIN 45 (ZPACK13415250) 500 ml Glass bottle for liquids DIN 45 (ZPACE8009063)
Packaging Information
spec_VWRC30110.264 spec_VWRC30110.264 spec_VWRC30110.297 spec_VWRC30110.297 spec_VWRC30110.320
Product specification
More
VWR® Volumetric Solutions for Titration
Product Literature
Frequently Bought Together