Perchloric acid 0.1 mol/l (0.1 N) in anhydrous acetic acid, Titripur® Reag. Ph. Eur., Reag. USP for titration in non-aqueous media, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7601-90-3
:
Formula:HClO₄
MDL Number:MFCD00011325
:100.46 g/mol
Density:1.06 g/cm³ (20 °C)
Flash Pt:40 °C
EINECS:200-580-7
UN:2789
ADR:8 (3),II
:M-Clarity™ quality level = MQ100
:GHS02
:GHS05
Storage Temperature:Ambient
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Form:liquid
- Amount-of-substance concentration:0.0995 - 0.1005
- Measurement uncertainty:+/- 0.0003
- Water content (according to Karl Fischer):0.1 - 0.2 %
- Traceability:NIST SRM
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.09065.1000_TDS_EN ProductPage_109065
Product specification
Merck Brighter titration Titripur® Merck Segment Brochure Merck Supelco® Titration goes Digital Merck Titration Guide for Titer Determination
Product Literature
msds_109065
SDS
Frequently Bought Together