Order Entry
Austria
ContactUsLinkComponent
Perchloric acid 0.1 mol/l (0.1 N) in anhydrous acetic acid
Flammable
Corrosive
Perchloric acid 0.1 mol/l (0.1 N) in anhydrous acetic acid
Catalog # 86732.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Corrosive
Perchloric acid 0.1 mol/l (0.1 N) in anhydrous acetic acid
Catalog # 86732.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.098 - 0.102 mol/l

Specification Test Results

About this item

 :
 :100.46 g/mol
Boiling Pt:120 °C (1013 hPa)
Density:1.050 g/cm³ (20 °C)
Flash Pt:40 °C (closed cup)
Storage Temperature:Ambient
EINECS:231-512-4
UN:2920
ADR:8 (3),II
 :GHS02
 :GHS05
CAS Number:7601-90-3
Formula:HClO₄
MDL Number:MFCD00011325