Order Entry
Austria
ContactUsLinkComponent
Hydrochloric acid concentrate for dilution to 1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Hydrochloric acid concentrate for dilution to 1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 32050.602
Supplier:  VWR Chemicals
CAS Number:  
undefined
Hydrochloric acid concentrate for dilution to 1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 32050.602
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    160 mL
  • Seq:
    2510

Specifications

Specification Test Results

  • Titer (20°C):
    0.998 - 1.002 mol/l

Specification Test Results

About this item

 :
ADR:8,III
 :Dose for preparation of 1 litre of solution.
 :36.46 g/mol
EINECS:231-595-7
UN:1789
 :GHS07
 :GHS05
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324