
Hydrochloric acid 1 mol/l (1 N), Reagent Grade
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
ADR:8,III
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Base Titration:900 – 1100 mmol/l
Specification Test Results
Frequently Bought Together