
Hydrochloric acid standardised solution 1.0 N
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
ADR:8,III
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS05
Storage Temperature:Ambient
Related Information
Frequently Bought Together