
Hydrochloric acid 0.5 mol/l (0.5 N), Titripur® Reag. USP volumetric solution, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
ADR:8,II
:M-Clarity™ quality level = MQ100
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS05
Storage Temperature:Ambient
Related Information
Certificates
Search by Lot NumberMerck Brighter titration Titripur® Merck Segment Brochure Merck Supelco® Smart Chemical Products
Product Literature
msds_109058
SDS
Frequently Bought Together