Order Entry
Austria
ContactUsLinkComponent
Hydrochloric acid 0.4 mol/l (0.4 N)
Corrosive
Hydrochloric acid 0.4 mol/l (0.4 N)
Catalog # 30021.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Hydrochloric acid 0.4 mol/l (0.4 N)
Catalog # 30021.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
Density:1.02 g/cm³
Storage Temperature:Ambient
EINECS:231-595-7
ADR:8,III
 :36.46 g/mol
UN:1789
 :GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324