Order Entry
Austria
ContactUsLinkComponent
 
Hydrochloric acid 0.2 mol/l (0.2 N), BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7651.9010
Supplier:  Avantor
CAS Number:  
Hydrochloric acid 0.2 mol/l (0.2 N), BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7651.9010
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    10 L
  • Seq:
    5214

Specifications

Specification Test Results

  • Titer (mol/l):
    0.1990 - 0.2010

Specification Test Results

About this item

 :
Density:1.004 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-595-7
ADR:8,III
 :36.46 g/mol
UN:1789
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324