Order Entry
Austria
ContactUsLinkComponent
Hydrochloric acid standardised solution 0.1 N
Hydrochloric acid standardised solution 0.1 N
Catalog # 35644.K2
CAS Number:  
Hydrochloric acid standardised solution 0.1 N
Catalog # 35644.K2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

CAS Number:7647-01-0
 :
Formula:HCl
MDL Number:MFCD00011324
Density:1.004 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8,III
 :36.46 g/mol