Order Entry
Austria
ContactUsLinkComponent
Hydrochloric acid standard solution 0.1 mol/l (0.1 N) in water
Corrosive
Hydrochloric acid standard solution 0.1 mol/l (0.1 N) in water
Certificates
undefined
Corrosive
Hydrochloric acid standard solution 0.1 mol/l (0.1 N) in water

Specification Test Results

  • Appearance:
    Clear colorless Solution
  • Strength:
    0.0998 to 0.1002 N (20°C)
  • Titration Iodometric:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together