Order Entry
Austria
ContactUsLinkComponent
Hydrochloric acid 0.02 mol/l (0.02 N), AVS TITRINORM volumetric solution
Hydrochloric acid 0.02 mol/l (0.02 N), AVS TITRINORM volumetric solution
Catalog # 98052.296
Supplier:  VWR Chemicals
CAS Number:  
undefined
Hydrochloric acid 0.02 mol/l (0.02 N), AVS TITRINORM volumetric solution
Catalog # 98052.296
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.01996 - 0.02004 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
ADR:8,III
 :36.46 g/mol
UN:1789
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324