Order Entry
Austria
ContactUsLinkComponent
Silver nitrate concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Corrosive
Environment
Silver nitrate concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 32056.602
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Environment
Silver nitrate concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 32056.602
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    60 mL
  • Seq:
    2385

Specifications

Specification Test Results

  • Titer (20°C):
    0.0995 to 0.1005 mol/l
  • Conforms to BDH 18041:
    Passes test

Specification Test Results

About this item

CAS Number:7761-88-8
 :
Formula:AgNO₃
MDL Number:MFCD00003414
 :169.87 g/mol
ADR:8,II
 :Dose for preparation of 1 litre of solution.
EINECS:231-853-9
UN:3264
 :GHS05
 :GHS09
Storage Temperature:Ambient