Order Entry
Austria
ContactUsLinkComponent
Sodium thiosulphate 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Sodium thiosulphate 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Supplier:  Merck
Certificates
Sodium thiosulphate 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.22 g/cm3 (20 °C)
  • pH:
    9 - 10 (H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together