Order Entry
Austria
ContactUsLinkComponent
Sodium hydroxide 0.1 mol concentrated aqueous solution 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Sodium hydroxide 0.1 mol concentrated aqueous solution 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Supplier:  Merck
Certificates
Sodium hydroxide 0.1 mol concentrated aqueous solution 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Supplier:  Merck
Certificates
Limited Time Sale
Select items below are currently on sale

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.09 g/cm3 (20 °C)
  • pH:
    13.8 (H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

Related Information

Frequently Bought Together