Order Entry
Austria
ContactUsLinkComponent
Nitric acid concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Corrosive
Nitric acid concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 32069.606
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Nitric acid concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 32069.606
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    60 mL
  • Seq:
    2385

Specifications

Specification Test Results

  • Titer (20°C):
    0.0998 - 0.1002 mol/l

Specification Test Results

About this item

 :
 :63.01 g/mol
Density:1.38 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2031
ADR:8,II
 :Dose for preparation of 1 litre of solution.
EINECS:231-714-2
 :GHS05
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349