Order Entry
Austria
ContactUsLinkComponent
Hydrochloric acid 0.5 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Corrosive
Irritant
Hydrochloric acid 0.5 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Catalog # 1.09971.0001
Supplier:  Merck
CAS Number:  
Corrosive
Irritant
Hydrochloric acid 0.5 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Catalog # 1.09971.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • Seq:
    0501

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.1 g/cm3 (20 °C)
  • pH:
     - 1 (H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

About this item

 :
ADR:8,
 :M-Clarity™ quality level = MQ100
 :36.46 g/mol
EINECS:231-595-7
UN:1789
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324