Hydrochloric acid 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8,
:M-Clarity™ quality level = MQ100
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS05
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Solubility:(20 °C) soluble
- Density:1.03 g/cm3 (20 °C)
- pH:- 1 (H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.09973.0001_TDS_EN ProductPage_109973 ProductPage_109973
Product specification
Merck Brighter titration Titripur®
Product Literature
msds_109973
SDS
Frequently Bought Together