

(±)-1-Bromo-2-methylbutane ≥97% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5973-11-5
:
Formula:C₅H₁₁Br
:151.05 g/mol
Merck Index:14,601
Boiling Pt:121…122 °C
Density:1.223
Flash Pt:22 °C (79 °F)
Storage Temperature:Ambient
EINECS:208-583-5
ADR:3,III
:Stabilised with potassium carbonate
UN:1993
:GHS02
:GHS07
Related Information
Frequently Bought Together