Order Entry
Austria
ContactUsLinkComponent
 
 
 
 
 
(±)-Epichlorhydrin for synthesis, Sigma-Aldrich®
 :  Merck
 
 
 
 
 
(±)-Epichlorhydrin for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    60 g/l (20 °C)
  • Melting Point:
    -57.2 °C
  • Molar Mass:
    92.52 g/mol
  • Boiling Point:
    116.5 °C (1013 hPa)
  • Vapor Pressure:
    16.5 hPa (20 °C)
  • Flash Point:
    28 °C
  • Refractive Index:
    1.4375 (20 °C, 589 nm)
  • Explosion Limit:
    2.3 - 34.4 %(V)
  • Density:
    1.18 g/cm3 (20 °C)
  • Ignition Point:
    416 °C
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic Materials

 

Frequently Bought Together