Order Entry
Austria
ContactUsLinkComponent
Sulphuric acid ≥90% for milk analysis
Corrosive
Sulphuric acid ≥90% for milk analysis
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
Sulphuric acid ≥90% for milk analysis
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 90 %
  • Density (20/4):
    1.815 - 1.825

Specification Test Results

Frequently Bought Together