Order Entry
Austria
ContactUsLinkComponent
Sulphuric acid 62% for the determination of fats in cheese according to Gerber
Sulphuric acid 62% for the determination of fats in cheese according to Gerber
Supplier:  VWR Chemicals
Certificates
undefined
Sulphuric acid 62% for the determination of fats in cheese according to Gerber
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Density (20/4):
    1.517 - 1.527
  • Assay:
    61.7 - 62.7 %

Specification Test Results

Frequently Bought Together