Order Entry
Austria
ContactUsLinkComponent
 
 
Sulphuric acid 25%, GPR RECTAPUR®
 
undefined
 
Sulphuric acid 25%, GPR RECTAPUR®
 

 

  • Assay:
    24 - 26 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Ignition residue:
    Max. 50 ppm
  • Cl (Chloride):
    Max. 5 ppm
  • Fe (Iron):
    Max. 5 ppm

 

Frequently Bought Together