Order Entry
Austria
ContactUsLinkComponent
Perrhenic acid 99.99% (trace metals basis) in aqueous solution
Corrosive
Irritant
Perrhenic acid 99.99% (trace metals basis) in aqueous solution
Certificates
undefined
Corrosive
Irritant
Perrhenic acid 99.99% (trace metals basis) in aqueous solution

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Solution
  • Strength:
    70 to 76.5 % (HReO4)
  • Total trace metal impurities:
    ≤100 ppm

Specification Test Results

Related Information

Frequently Bought Together