Orders LinkContactUsLinkComponent
Sample containers, snap-seal, Corning®
Supplier:   Corning
Sample containers, snap-seal, Corning®
Supplier:  Corning
Certificates
Sample containers, snap-seal, Corning®
Sample containers, snap-seal, Corning®
5 Options Available Below
Product Details & Documents
PP, disposable, translucent.
  • Snap-Seal locking device keeps the cap closed and secure
  • Hinged cap reduces the chance of sample contamination
  • Graduations in both milliliters and ounces
  • Rough source for marking
These wide mouth sample containers are designed for a wide variety of applications and provide a reliable leak-tight seal when closed properly.
 : The product(s) on this page are outside the scope of the EU Directive 2019/904.
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Cap colour
    Packed
    Material
    Availability
    Price
  • 45 ml
    30 mm
    84 mm
    Natural
    400/pack
    PP
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 160,00 €
    160,00 €
    1 * 400 ST
       160,00 €
  • 120 ml
    45 mm
    91 mm
    Natural
    200/pack
    PP
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 106,00 €
    106,00 €
    1 * 200 ST
       106,00 €
  • 120 ml
    68 mm
    52 mm
    Natural
    200/pack
    PP
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 109,00 €
    109,00 €
    1 * 200 ST
       109,00 €
  • 240 ml
    80 mm
    75 mm
    Natural
    100/pack
    PP
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 78,20 €
    78,20 €
    1 * 100 ST
       78,20 €
  • 300 ml
    63 mm
    112 mm
    Natural
    100/pack
    PP
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 89,10 €
    89,10 €
    1 * 100 ST
       89,10 €