Orders LinkContactUsLinkComponent
Jars, multipurpose, with covers, Nalgene®
Jars, multipurpose, with covers, Nalgene®
   Sustainable Options Available
Jars, multipurpose, with covers, Nalgene®
   Sustainable Options Available
Certificates
Jars, multipurpose, with covers, Nalgene®
3 Options Available Below
Product Details & Documents
Polycarbonate, clear.
  • Drill, tap or apply clamps to jars to attach heating or cooling equipment
  • Very durable
  • Autoclavable
Multipurpose jars with cover, which can be used as water baths, aquaria/terraria or freeze-drying chambers.
Specifications for Product Family
Specifications
  • Catalog No:
  • 216-8262
  • 216-8264
  • 216-8265
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Availability
    Price
  • 2200 ml
    133 mm
    195 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 108,00 €
    108,00 €
    1 * 1 ST
       108,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-8262
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 4700 ml
    170 mm
    239 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 140,00 €
    140,00 €
    1 * 1 ST
       140,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-8264
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 8300 ml
    222 mm
    258 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 222,00 €
    222,00 €
    1 * 1 ST
       222,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-8265
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact