Order Entry
Austria
ContactUsLinkComponent
Accessories for graduated pipettes BLAUBRAND®
Accessories for graduated pipettes BLAUBRAND®
Catalog # 612-1199
Supplier:  Brand
undefined
Accessories for graduated pipettes BLAUBRAND®
Catalog # 612-1199
Supplier:  Brand

Specifications

  • Description:
    Cotton roving for pipettes, cotton wool, degreased, 1 kg
  • Pk:
    1

Specifications

About this item

These plugs made of 100% cotton wool may prolong delivery time and may therefore influence the accuracy of measurement.

In tests using pipettes that have had cotton plugs inserted by a cotton plugging machine, the results were within the indicated error limits.

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904