VWR® Volumetric Flasks, Class A, Glass, Amber, NS Plastic Stopper
Borosilicate 3.3 glass, amber, class A.
- White PE stopper
- Marks and inscriptions in white enamel
- Calibrated to contain (TC, In)
DIN EN ISO 1042
: Batch number and batch certificate included.
Frequently Bought Together