Order Entry
Austria
ContactUsLinkComponent
Sodium hydroxide 50% in aqueous solution, EMSURE® for analysis, Supelco®
Corrosive
Sodium hydroxide 50% in aqueous solution, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates
Corrosive
Sodium hydroxide 50% in aqueous solution, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (acidimetric, as NaOH):
    49.0 - 51.0 %
  • Carbonate (as Na₂CO₃):
    ≤ 0.2
  • Color (Hazen):
    ≤ 10 Hazen
  • Chloride (Cl):
    ≤ 0.01 %
  • Phosphate (PO₄):
    ≤ 0.0005 %
  • Silicate (SiO₂):
    ≤ 0.005 %
  • Sulfate (SO₄):
    ≤ 0.005 %
  • Total nitrogen (N):
    ≤ 0.0003 %
  • Heavy metals (as Pb):
    ≤ 0.0005 %
  • Al (Aluminium):
    ≤ 0.0005 %
  • Fe (Iron):
    ≤ 0.0005 %
  • K (Potassium):
    ≤ 0.01 %

Specification Test Results

Frequently Bought Together