Order Entry
Austria
ContactUsLinkComponent
Sodium hydroxide 40% in aqueous solution, BAKER, Laboratory Reagent, J.T.Baker®
Corrosive
Sodium hydroxide 40% in aqueous solution, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 6165.9010
Supplier:  Avantor
CAS Number:  
Corrosive
Sodium hydroxide 40% in aqueous solution, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 6165.9010
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    10 L
  • Seq:
    5214

Specifications

Specification Test Results

  • Assay:
    min. 40.0 %
  • Density (g/ml) at 20°C:
    min. 1.43
  • Iron (Fe):
    max. 5
  • Nitrogen Compounds (as N):
    max. 5

Specification Test Results

About this item

 :
 :40 g/mol
Boiling Pt:119…135 °C (1013 hPa)
Melting Pt:9…10 °C
Density:1,53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548