Order Entry
Austria
ContactUsLinkComponent
 
 
Sodium hydroxide 32% in aqueous solution, AnalaR NORMAPUR® analytical reagent, for nitrogen analysis
 
undefined
 
Sodium hydroxide 32% in aqueous solution, AnalaR NORMAPUR® analytical reagent, for nitrogen analysis
 

 

  • Assay:
    Min. 32 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Total N (Nitrogen):
    Max. 1 ppm
  • Cl (Chloride):
    Max. 20 ppm
  • CO3 (as Na2CO3):
    Max. 1.0 %
  • SiO4 (as SiO2):
    Max. 50 ppm
  • SO4 (Sulphate):
    Max. 30 ppm
  • Al (Aluminium):
    Max. 10 ppm
  • Fe (Iron):
    Max. 10 ppm

 

Frequently Bought Together