
Sodium hydroxide 33% (w/w) in water, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:119…135 °C (1013 hPa)
Melting Pt:8 °C
Density:1.36 g/cm³
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Clear colorless Solution
- Heavy metals:≤10 ppm
- Density:(20°C) 1.355 to 1.365 kg/l
- Titration with HCl:32.5 to 33.5 %
- Sodium carbonate (Na2CO3):≤0.1 % (Na2CO3)
- Sodium sulfate (Na2SO4):≤50 ppm (Na2SO4)
- Sodium chloride (NaCl):≤50 ppm (NaCl)
Specification Test Results
Frequently Bought Together