Order Entry
Austria
ContactUsLinkComponent
Sodium hydroxide 30% (w/w) in aqueous solution, AnalaR NORMAPUR® analytical reagent
Sodium hydroxide 30% (w/w) in aqueous solution, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Sodium hydroxide 30% (w/w) in aqueous solution, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    30 - 31 %
  • Total alkalies:
    30.0 to 31.0 %
  • Density (20/4):
    1.32 - 1.34
  • Heavy metals (as Pb):
    Max. 2 ppm
  • Silica:
    Max. 5 ppm
  • Total N (Nitrogen):
    Max. 10 ppm
  • Cl (Chloride):
    Max. 200 ppm
  • CO3 (as Na2CO3):
    Max. 1 %
  • PO4 (Phosphate):
    Max. 1 ppm
  • SO4 (Sulphate):
    Max. 10 ppm
  • Al (Aluminium):
    Max. 2 ppm
  • Ba (Barium):
    Max. 2 ppm
  • Ca (Calcium):
    Max. 2 ppm
  • Cd (Cadmium):
    Max. 0.1 ppm
  • Co (Cobalt):
    Max. 0.3 ppm
  • Cr (Chromium):
    Max. 2 ppm
  • Cu (Copper):
    Max. 0.3 ppm
  • Fe (Iron):
    Max. 2 ppm
  • Hg (Mercury):
    Max. 0.02 ppm
  • K (Potassium):
    Max. 50 ppm
  • Mg (Magnesium):
    Max. 2 ppm
  • Mn (Manganese):
    Max. 0.1 ppm
  • Ni (Nickel):
    Max. 0.6 ppm
  • Pb (Lead):
    Max. 0.1 ppm
  • Zn (Zinc):
    Max. 0.3 ppm

Specification Test Results

Frequently Bought Together