


Nitric acid 69%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7697-37-2
:
Formula:HNO₃
MDL Number:MFCD00011349
:63.01 g/mol
Merck Index:13,06608
Boiling Pt:120.5 °C (1013 hPa)
Melting Pt:–42 °C
Density:1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8 (5.1),II
:GHS03
:GHS05
:GHS06
Specification Test Results
- Assay:69.0 - 70.0 %
- Appearance of solution:Passes test Ph.Eur.
- Identification (NO3):Passes test Ph.Eur.
- Colouration:Max. 10 APHA
- Density (20/20):1.384 - 1.416
- Heavy metals (as Pb):Max. 0.2 ppm
- Ignition residue (SO4):Max. 5 ppm
- Silica:Max. 0.2 ppm
- Cl (Chloride):Max. 0.5 ppm
- PO4 (Phosphate):Max. 2 ppm
- SO4 (Sulphate):Max. 1 ppm
- Al (Aluminium):Max. 0.05 ppm
- As (Arsenic):Max. 0.01 ppm
- Ba (Barium):Max. 0.02 ppm
- Ca (Calcium):Max. 0.1 ppm
- Cd (Cadmium):Max. 0.01 ppm
- Co (Cobalt):Max. 0.01 ppm
- Cr (Chromium):Max. 0.02 ppm
- Cu (Copper):Max. 0.01 ppm
- Fe (Iron):Max. 0.1 ppm
- K (Potassium):Max. 0.05 ppm
- Mg (Magnesium):Max. 0.03 ppm
- Mn (Manganese):Max. 0.01 ppm
- Mo (Molybdenum):Max. 0.02 ppm
- Na (Sodium):Max. 0.3 ppm
- Ni (Nickel):Max. 0.01 ppm
- Pb (Lead):Max. 0.01 ppm
- Sr (Strontium):Max. 0.01 ppm
- Zn (Zinc):Max. 0.05 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 1L Glass bottle Safebreak for liquids DIN 45 (ZPACK13415111) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 2,5L Glass bottle Safebreak for liquids DIN 45 (ZPACK13415250)
Packaging Information
SDS_EN_CH_VWRC20425.pdf SDS_EN_DE_VWRC20425.pdf SDS_EN_NL_VWRC20425.pdf SDS_EN_UK_VWRC20425.pdf sds_VWRC20425
SDS
More
spec_VWRC20425.242 spec_VWRC20425.242 spec_VWRC20425.297 spec_VWRC20425.297 spec_VWRC20425.322
Product specification
More
Frequently Bought Together