Order Entry
Austria
ContactUsLinkComponent
Nitric acid 69%, EMPARTA® ACS analytical reagent, Supelco®
Oxidizing
Toxic
Corrosive
Nitric acid 69%, EMPARTA® ACS analytical reagent, Supelco®
Catalog # 1.01832.2500
Supplier:  Merck
CAS Number:  
Oxidizing
Toxic
Corrosive
Nitric acid 69%, EMPARTA® ACS analytical reagent, Supelco®
Catalog # 1.01832.2500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -41 °C
  • Boiling Point:
    122 °C (1013 hPa)
  • Vapor Pressure:
    9.4 hPa (20 °C)
  • Density:
    1.41 g/cm3 (20 °C)
  • pH:
     - 1 (H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive material

Specification Test Results

About this item

 :
 :63.01 g/mol
Merck Index:13,06608
Boiling Pt:120.5 °C (1013 hPa)
Melting Pt:–42 °C
Density:1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8 (5.1),II
 :M-Clarity™ quality level = MQ200
 :GHS03
 :GHS06
 :GHS05
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349