Order Entry
Austria
ContactUsLinkComponent
Nitric acid 69,0 - 70,0%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®
Oxidizing
Corrosive
Toxic
Nitric acid 69,0 - 70,0%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Oxidizing
Corrosive
Toxic
Nitric acid 69,0 - 70,0%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    69.0 - 70.0 %
  • ACS - Assay (HNO₃):
    69.0 - 70.0 %
  • Appearance:
    Passes Test
  • ACS - Color (APHA):
    ≤ 10
  • ACS - Residue after Ignition:
    ≤ 2 ppm
  • Chloride (Cl):
    ≤ 0.04 ppm
  • Phosphate (PO₄):
    ≤ 0.10 ppm
  • Sulfate (SO₄):
    ≤ 0.4 ppm
  • Trace Impurities - Aluminum (Al):
    ≤ 30.0 ppb
  • Arsenic and Antimony (as As):
    ≤ 5.0 ppb
  • Trace Impurities - Barium (Ba):
    ≤ 1.0 ppb
  • Trace Impurities - Beryllium (Be):
    ≤ 1.0 ppb
  • Trace Impurities - Bismuth (Bi):
    ≤ 1.0 ppb
  • Trace Impurities - Boron (B):
    ≤ 4.0 ppb
  • Trace Impurities - Cadmium (Cd):
    ≤ 1.0 ppb
  • Trace Impurities - Calcium (Ca):
    ≤ 50.0 ppb
  • Trace Impurities - Chromium (Cr):
    ≤ 10.0 ppb
  • Trace Impurities - Cobalt (Co):
    ≤ 1.0 ppb
  • Trace Impurities - Copper (Cu):
    ≤ 1.0 ppb
  • Trace Impurities - Gallium (Ga):
    ≤ 20.0 ppb
  • Trace Impurities - Germanium (Ge):
    ≤ 4.0 ppb
  • Trace Impurities - Gold (Au):
    ≤ 4.0 ppb
  • Heavy Metals (as Pb):
    ≤ 100 ppb
  • Trace Impurities - Iron (Fe):
    ≤ 10.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤ 0.5 ppb
  • Trace Impurities - Lithium (Li):
    ≤ 1.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤ 7.0 ppb
  • Trace Impurities - Manganese (Mn):
    ≤ 1.0 ppb
  • Trace Impurities - Mercury (Hg):
    ≤ 0.5 ppb
  • Trace Impurities - Molybdenum (Mo):
    ≤ 5.0 ppb
  • Trace Impurities - Nickel (Ni):
    ≤ 1.0 ppb
  • Trace Impurities - Niobium (Nb):
    ≤ 1.0 ppb
  • Trace Impurities - Potassium (K):
    ≤ 5.0 ppb
  • Trace Impurities - Silicon (Si):
    ≤ 20.0 ppb
  • Trace Impurities - Silver (Ag):
    ≤ 1.0 ppb
  • Trace Impurities - Sodium (Na):
    ≤ 200.0 ppb
  • Trace Impurities - Strontium (Sr):
    ≤ 1.0 ppb
  • Trace Impurities - Tantalum (Ta):
    ≤ 2.0 ppb
  • Trace Impurities - Thallium (Tl):
    ≤ 5.0 ppb
  • Trace Impurities - Tin (Sn):
    ≤ 5.0 ppb
  • Trace Impurities - Vanadium (V):
    ≤ 1.0 ppb
  • Trace Impurities - Zinc (Zn):
    ≤ 5.0 ppb
  • Trace Impurities - Zirconium (Zr):
    ≤ 1.0 ppb

Specification Test Results

Related Information

Frequently Bought Together