


Nitric acid 65%, EMSURE® Reag. Ph. Eur., ISO for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7697-37-2
:
Formula:HNO₃
MDL Number:MFCD00011349
:63.01 g/mol
Merck Index:13,06608
Boiling Pt:121 °C (1013 hPa)
Melting Pt:–32 °C
Density:1.39 g/cm3 (20 °C)
Storage Temperature:Ambient
UN:2031
ADR:8 (5.1),II
:M-Clarity™ quality level = MQ300
EINECS:231-714-2
:GHS03
:GHS06
:GHS05
Specification Test Results
- Assay (alkalimetric):≥ 65.0 %
- Identity:passes test
- Appearance:passes test
- Color (Hazen):≤ 10 Hazen
- Density (d 20/20):1.384 - 1.416
- Chloride (Cl):≤ 0.2 ppm
- Phosphate (PO₄):≤ 0.2 ppm
- Sulfate (SO₄):≤ 0.5 ppm
- Heavy metals (as Pb):≤ 0.2 ppm
- Ag (Silver):≤ 0.010 ppm
- Al (Aluminium):≤ 0.050 ppm
- As (Arsenic):≤ 0.010 ppm
- Au (Gold):≤ 0.050 ppm
- Ba (Barium):≤ 0.010 ppm
- Be (Beryllium):≤ 0.010 ppm
- Bi (Bismuth):≤ 0.020 ppm
- Ca (Calcium):≤ 0.100 ppm
- Cd (Cadmium):≤ 0.010 ppm
- Co (Cobalt):≤ 0.010 ppm
- Cr (Chromium):≤ 0.020 ppm
- Cu (Copper):≤ 0.010 ppm
- Fe (Iron):≤ 0.100 ppm
- Ga (Gallium):≤ 0.050 ppm
- Ge (Germanium):≤ 0.020 ppm
- In (Indium):≤ 0.020 ppm
- K (Potassium):≤ 0.100 ppm
- Li (Lithium):≤ 0.010 ppm
- Mg (Magnesium):≤ 0.050 ppm
- Mn (Manganese):≤ 0.010 ppm
- Mo (Molybdenum):≤ 0.010 ppm
- Na (Sodium):≤ 0.200 ppm
- Ni (Nickel):≤ 0.020 ppm
- Pb (Lead):≤ 0.010 ppm
- Pt (Platinum):≤ 0.100 ppm
- Sr (Strontium):≤ 0.010 ppm
- Ti (Titanium):≤ 0.020 ppm
- Tl (Thallium):≤ 0.020 ppm
- V (Vanadium):≤ 0.010 ppm
- Zn (Zinc):≤ 0.020 ppm
- Zr (Zirconium):≤ 0.020 ppm
- Residue on ignition (as SO₄):≤ 3 ppm
- Tested by the dithizone:passes test
Specification Test Results
Related Information
Frequently Bought Together