Order Entry
Austria
ContactUsLinkComponent
Hydrofluoric acid 47 - 51%, TraceSELECT™ for trace analysis, Fluka™
Corrosive
Toxic
Hydrofluoric acid 47 - 51%, TraceSELECT™ for trace analysis, Fluka™
Catalog # 47559-500ML
CAS Number:  
Corrosive
Toxic
Hydrofluoric acid 47 - 51%, TraceSELECT™ for trace analysis, Fluka™
Catalog # 47559-500ML
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Assay:
    47.0 - 51.0 %
  • Density (D 20/4):
    1.17
  • Silver (Ag):
    Max. 0.001 ppm
  • Aluminium (Al):
    Max. 0.005 ppm
  • Arsenic (As):
    Max. 0.005 ppm
  • Gold (Au):
    Max. 0.001 ppm
  • Barium (Ba):
    Max. 0.001 ppm
  • Beryllium (Be):
    Max. 0.001 ppm
  • Bismuth (Bi):
    Max. 0.001 ppm
  • Calcium (Ca):
    Max. 0.01 ppm
  • Cadmium (Cd):
    Max. 0.001 ppm
  • Cesium (Cs):
    Max. 0.001 ppm
  • Cobalt (Co):
    Max. 0.001 ppm
  • Chromium (Cr):
    Max. 0.001 ppm
  • Copper (Cu):
    Max. 0.001 ppm
  • Iron (Fe):
    Max. 0.01 ppm
  • Gallium (Ga):
    Max. 0.001 ppm
  • Germanium (Ge):
    Max. 0.001 ppm
  • Mercury (Hg):
    Max. 0.005 ppm
  • Indium (In):
    Max. 0.001 ppm
  • Potassium (K):
    Max. 0.01 ppm
  • Lithium (Li):
    Max. 0.001 ppm
  • Magnesium (Mg):
    Max. 0.005 ppm
  • Manganese (Mn):
    Max. 0.001 ppm
  • Molybdenum (Mo):
    Max. 0.001 ppm
  • Sodium (Na):
    Max. 0.01 ppm
  • Nickel (Ni):
    Max. 0.001 ppm
  • Lead (Pb):
    Max. 0.002 ppm
  • Rubidium (Rb):
    Max. 0.001 ppm
  • Selenium (Se):
    Max. 0.01 ppm
  • Tin (Sn):
    Max. 0.001 ppm
  • Strontium (Sr):
    Max. 0.001 ppm
  • Titanium (Ti):
    Max. 0.005 ppm
  • Thallium (Tl):
    Max. 0.001 ppm
  • Vanadium (V):
    Max. 0.001 ppm
  • Zinc (Zn):
    Max. 0.002 ppm
  • Zirconium (Zr):
    Max. 0.001 ppm
  • Vapor Density:
    1.27 (vs air)

Specification Test Results

About this item

 :
 :20.01 g/mol
Boiling Pt:105 °C (1.013 hPa)
Melting Pt:–35 °C
Density:1.170 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-634-8
UN:1790
ADR:8,II
 :GHS05
 :GHS06
CAS Number:7664-39-3
Formula:HF
MDL Number:MFCD00011346