Hydrofluoric acid 40 - 45%, Technical Grade, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7664-39-3
:
Formula:HF
MDL Number:MFCD00011346
Boiling Pt:110 °C (1.013 hPa)
Melting Pt:–40 °C
Density:1.140 g/cm³ (20 °C)
Molecular Weight:20.01 g/mol
Storage Temperature:Ambient
EINECS:231-634-8
UN:1790
ADR:8 II
:GHS06
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Vapor Density:1.27 (vs air)
- Assay:40 - 45 %
- Hexafluorosilicic acid (H₂SiF₆):Max. 0.02 %
- Sulfate (SO₄):Max. 0.01 %
- Vapor Pressure:25 mmHg ( 20 °C)
Specification Test Results
Frequently Bought Together