Order Entry
Austria
ContactUsLinkComponent
 
 
 
Perchloric acid ≥70%, GPR RECTAPUR®
  20587.296
 :  
undefined
 
 
Perchloric acid ≥70%, GPR RECTAPUR®
  20587.296
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Assay:
    69 - 72 %
  • Ignition residue (SO4):
    Max. 0.02 %
  • Cl (Chloride):
    Max. 25 ppm
  • Fe (Iron):
    Max. 10 ppm

 

 

 :
 :100.46 g/mol
 :160…181 °C (1013 hPa)
 :–112,2 °C
 :1.53…1.768 g/cm³ (20 °C)
 :Ambient
 :231-512-4
 :1873
 :5.1 (8),I
 :GHS03
 :GHS05
 :7601-90-3
 :HClO₄
 :MFCD00011325