Order Entry
Austria
ContactUsLinkComponent
Labels, hazard label set for self labelling acc. to GHS
Labels, hazard label set for self labelling acc. to GHS
Catalog # 1.00801.0001
Supplier:  Merck
undefined
Labels, hazard label set for self labelling acc. to GHS
Catalog # 1.00801.0001
Supplier:  Merck

Specifications

  • Colour:
    White/black/red
  • Description:
    GHS labels
  • Pk:
    1

Specifications

About this item

With implementation of CLP/EU-GHS (Global Harmonised System) directive in January 2009, the procedures for the classification, labelling and packing of hazardous chemicals have been redefined.

  • Works according to this directive
  • Customer's individual mixtures can also be labelled according to GHS, GLP guidelines and DIN EN ISO
Packaging: Set includes 20 adhesive labels 80×110 mm, 168 adhesive pictograms, 40 adhesive signal words and a pack insert with H, P and EUH statements.